Indium(III) hydroxide ≥99.5%

Supplier: BeanTown Chemical

145970-10G 145970-50G
BT145970-10GEA 63 USD
BT145970-10G BT145970-50G
Indium(III) hydroxide ≥99.5%
Indium(III) hydroxide

CAS: 20661-21-6; EC No: 243-947-7; MDL No: MFCD00011056 Powder; Linear Formula: In(OH)3; MW: 165.84 Melting Point: 150° Density (g/mL): 4.45


  • Powder

Formula: H₃InO₃
Melting Pt: 150 °C
Density: 4.45 g/mL
MDL Number: MFCD00011056
CAS Number: 20661-21-6
EINECS: 243-947-7

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR