Cobalt(II) stearate
Supplier: BeanTown Chemical
Synonyms:
Cobalt(II) Stearate, Cobalt stearate
145960-1KG
145960-250G
145960-50G
BT145960-1KGEA
125
USD
BT145960-1KG
BT145960-250G
BT145960-50G
Cobalt(II) stearate
Cobalt(II) stearate
CAS: 1002-88-6; EC No: 213-694-7; MDL No: MFCD00054343; RTECS: RG1212000
Pellets; Molecular Formula: C36H70CoO4 ; MW: 625.89
- RTECS: RG1212000
- Pellets
Formula:
C₃₆H₇₀CoO₄ MW: 625.872 g/mol |
MDL Number:
MFCD00054343 CAS Number: 1002-88-6 EINECS: 213-694-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...