5-(Chloromethyl)uracil ≥97%

Supplier: BeanTown Chemical

Synonyms: 5-(Chloromethyl)pyrimidine-2,4(1H,3H)-dione

145935-1G 145935-250MG 145935-5G
BT145935-1GEA 124 USD
BT145935-1G BT145935-250MG BT145935-5G
5-(Chloromethyl)uracil ≥97%
5-(Chloromethyl)uracil

CAS: 3590-48-5; MDL No: MFCD00218445 Powder; Molecular Formula: C5H5ClN2O2; MW: 160.56 Melting Point: 350°


  • Powder

Formula: C₅H₅ClN₂O₂
MW: 160.56 g/mol
Melting Pt: >350 °C
MDL Number: MFCD00218445
CAS Number: 3590-48-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR