Chloranilic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
Chloranilicacid
145900-100G
145900-25G
145900-500G
BT145900-100GEA
105
USD
BT145900-100G
BT145900-25G
BT145900-500G
Chloranilic acid ≥98%
Chloranilic acid
CAS: 87-88-7; EC No: 201-780-7; MDL No: MFCD00001596; RTECS: DK4005000
Crystalline/Powder; Molecular Formula: C6H2Cl2O4; MW: 208.99
Melting Point:
- RTECS: DK4005000
- Crystalline/Powder
Formula:
C₆H₂Cl₂O₄ MW: 208.98 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00001596 CAS Number: 87-88-7 EINECS: 201-780-7 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...