Mercury (II) sulphide, black ≥99.999%

Supplier: BeanTown Chemical

145885-25G 145885-5G
BT145885-25GEA 778 USD
BT145885-25G BT145885-5G
Mercury (II) sulphide, black ≥99.999%
Mercury (II) sulphide

CAS: 1344-48-5; EC No: 215-696-3; MDL No: MFCD00011046; RTECS: OX0720000 Powder; Molecular Formula: HgS; MW: 232.66 Melting Point: 584° (sublimes) Density (g/mL): 7.7


  • RTECS: OX0720000
  • Powder

Formula: HgS
MW: 232.66 g/mol
Melting Pt: 584 °C (subl.)
Density: 7.7 g/mL
MDL Number: MFCD00011046
CAS Number: 1344-48-5
EINECS: 215-696-3
UN: 2025
ADR: 6.1 II

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR