2-Amino-5-nitropyridine ≥98%

Supplier: BeanTown Chemical

Synonyms: 5-Nitropyridin-2-ylamine, 5-Nitropyridin-2-amine, 5-Nitro-2-pyridylamine

145875-100G 145875-25G 145875-500G
BT145875-100GEA 487 USD
BT145875-100G BT145875-25G BT145875-500G
2-Amino-5-nitropyridine ≥98%
2-Amino-5-nitropyridine

CAS: 4214-76-0; EC No: 224-145-6; MDL No: MFCD00006325 Powder; Molecular Formula: C5H5N3O2 ; MW: 139.11 Melting Point: 186-188° Light Sensitive


  • Light Sensitive
  • Powder

Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
Melting Pt: 186…188 °C
MDL Number: MFCD00006325
CAS Number: 4214-76-0
EINECS: 224-145-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR