2-Chloro-N-methoxy-N-methylacetamide ≥98%

Supplier: BeanTown Chemical

Synonyms: Methyl 2-Chloro-N-methylacetohydroxamate

145835-25G 145835-5G
BT145835-25GEA 231 USD
BT145835-25G BT145835-5G
2-Chloro-N-methoxy-N-methylacetamide ≥98%
2-Chloro-N-methoxy-N-methylacetamide

CAS: 67442-07-3; MDL No: MFCD00134232 Crystalline; Molecular Formula: C4H8ClNO2; MW: 137.57 Melting Point: 39-41°; Flash point: 104°C (219°F)


  • Crystalline

Formula: C₄H₈ClNO₂
MW: 137.56 g/mol
Melting Pt: 39…41 °C
Flash Pt: 104 °C (219 °F)
MDL Number: MFCD00134232
CAS Number: 67442-07-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR