Tris(trimethylsilyl)phosphite ≥96%

Supplier: BeanTown Chemical

Synonyms: Tris(trimethylsilyl)phosphite

145820-2G 145820-500MG
BT145820-2GEA 48 USD
BT145820-2G BT145820-500MG
Tris(trimethylsilyl)phosphite ≥96%
Tris(trimethylsilyl) phosphite

CAS: 1795-31-9; MDL No: MFCD00015588 Liquid; Linear Formula: [(CH3)3SiO]3P; MW: 298.54 Boiling Point: 78-81°/8 mmHg; Flash point: 110°C (230°F) Density (g/mL): 0.893; Refractive Index: 1.409 Air Sensitive, Moisture Sensitive


  • Refractive index: 1.409
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: C₉H₂₇O₃PSi₃
Boiling Pt: 78…81 °C/8 mmHg
Density: 0.893 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00015588
CAS Number: 1795-31-9

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR