Iron(II) sulphate heptahydrate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Iron vitriol, Ferrous sulphate heptahydrate
145775-10KG
145775-2.5KG
145775-500G
BT145775-10KGEA
290
USD
BT145775-10KG
BT145775-2.5KG
BT145775-500G
Iron(II) sulphate heptahydrate ≥98%
Iron(II) sulphate heptahydrate
CAS: 7782-63-0; EC No: 231-753-5; MDL No: MFCD00149719; RTECS: NO8510000
Crystalline; Linear Formula: FeSO4·7H2O; MW: 278.02
Density (g/mL): 1.898
Air Sensitive
- RTECS: NO8510000
- Air Sensitive
- Crystalline
Formula:
FeSO₄·7H₂O MW: 278.02 g/mol Melting Pt: 64 Density: 1.898 g/mL |
MDL Number:
MFCD00149719 CAS Number: 7782-63-0 EINECS: 231-753-5 Merck Index: 13,04091 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...