4-Chloro-7-methoxyquinoline

Supplier: BeanTown Chemical

145760-1G 145760-5G
BT145760-1GEA 74 USD
BT145760-1G BT145760-5G
4-Chloro-7-methoxyquinoline
4-Chloro-7-methoxyquinoline

CAS: 68500-37-8; MDL No: MFCD06659036 Solid; Molecular Formula: C10H8ClNO; MW: 193.63 Melting Point: 84-85°


  • Solid

Formula: C₁₀H₈ClNO
MW: 193.6300 g/mol
Melting Pt: 84…85 °C
MDL Number: MFCD06659036
CAS Number: 68500-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR