4-(Dimethylamino)benzaldehyde ≥99% ACS

Supplier: BeanTown Chemical

145745-100G 145745-25G 145745-500G
BT145745-100GEA 89.26 USD
BT145745-100G BT145745-25G BT145745-500G
4-(Dimethylamino)benzaldehyde ≥99% ACS
4-(Dimethylamino)benzaldehyde

CAS: 100-10-7; EC No: 202-819-0; MDL No: MFCD00003381; RTECS: CU5775000 Solid; Molecular Formula: C9H11NO; MW: 149.19 Melting Point: 72-75° Air Sensitive, Light Sensitive


  • RTECS: CU5775000
  • Air Sensitive, Light Sensitive
  • Solid

Formula: (CH₃)₂NC₆H₄CHO
Melting Pt: 72…75 °C
MDL Number: MFCD00003381
CAS Number: 100-10-7
EINECS: 202-819-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59709e90756a237e9c4bd808e8fd50ac9367d91&vid=909765f6b48eb3673cc290753e0ead3358b739cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR