4-Isopropoxybenzaldehyde ≥97%

Supplier: BeanTown Chemical

Synonyms: ALDH1A3-IN-3

145715-1G 145715-25G 145715-5G
BT145715-1GEA 35 USD
BT145715-1G BT145715-25G BT145715-5G
4-Isopropoxybenzaldehyde ≥97%
4-Isopropoxybenzaldehyde

CAS: 18962-05-5; EC No: 000-000-0; MDL No: MFCD00052357 Liquid; Molecular Formula: C10H12O; MW: 164.21 Boiling Point: 108-110°/5 mmHg Refractive Index: 1.547 Air Sensitive


  • Refractive index: 1.547
  • Air Sensitive
  • Liquid

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 108…110 °C/5 mmHg
MDL Number: MFCD00052357
CAS Number: 18962-05-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR