2-Fluorobenzoic acid ≥98%
Supplier: BeanTown Chemical
145710-100G
145710-500G
BT145710-100GEA
83
USD
BT145710-100G
BT145710-500G
2-Fluorobenzoic acid ≥98%
2-Fluorobenzoic acid
CAS: 445-29-4; EC No: 207-158-1; MDL No: MFCD00002405; RTECS: DH0250000
Powder; Molecular Formula: C7H5FO2; MW: 140.11
Melting Point: 122-125°
- RTECS: DH0250000
- Powder
Formula:
C₇H₅FO₂ Melting Pt: 122…125 °C |
MDL Number:
MFCD00002405 CAS Number: 445-29-4 EINECS: 207-158-1 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...