Lithium metaborate ≥99.995%

Supplier: BeanTown Chemical

145695-100G 145695-25G 145695-500G
BT145695-100GEA 242 USD
BT145695-100G BT145695-25G BT145695-500G
Lithium metaborate ≥99.995%
Lithium metaborate

CAS: 13453-69-5; EC No: 236-631-5; MDL No: MFCD00011089 Powder; Linear Formula: LiBO2; MW: 49.75 Melting Point: 845° Hygroscopic


  • Hygroscopic
  • Powder

Formula: BLiO₂
Melting Pt: 845 °C
MDL Number: MFCD00011089
CAS Number: 13453-69-5
EINECS: 236-631-5

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR