1-Chloro-4-iodobenzene ≥99%

Supplier: BeanTown Chemical

145680-100G 145680-25G
BT145680-100GEA 122 USD
BT145680-100G BT145680-25G
1-Chloro-4-iodobenzene ≥99%
1-Chloro-4-iodobenzene

CAS: 637-87-6; EC No: 211-305-5; MDL No: MFCD00001053 Crystalline; Molecular Formula: C6H4ClI; MW: 238.45 Melting Point: 53-54°; Boiling Point: 226-227°; Flash point: 108°C (226°F) Light Sensitive


  • Light Sensitive
  • Crystalline

Formula: C₆H₄ClI
MW: 238.45 g/mol
Boiling Pt: 226…227 °C
Melting Pt: 53…54 °C
Flash Pt: 108 °C (226 °F)
MDL Number: MFCD00001053
CAS Number: 637-87-6
EINECS: 211-305-5

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR