Pyromellitic dianhydride ≥97%
Supplier: BeanTown Chemical
Synonyms:
1,2,4,5-Benzenetetracarboxylic anhydride
145675-100G
145675-2.5KG
145675-500G
BT145675-100GEA
39
USD
BT145675-100G
BT145675-2.5KG
BT145675-500G
Pyromellitic dianhydride ≥97%
Pyromellitic dianhydride
CAS: 89-32-7; EC No: 201-898-9; MDL No: MFCD00005005; RTECS: DB9300000
Solid; Molecular Formula: C10H2O6; MW: 218.12
Melting Point: 283-286°; Boiling Point: 397-400°; Flash point: 380°C (716°F)
Density (g/mL): 1.68
Moisture Sensitive
- RTECS: DB9300000
- Moisture Sensitive
- Solid
Formula:
C₁₀H₂O₆ Boiling Pt: 397…400 °C Melting Pt: 283…286 °C Density: 1.68 g/mL Flash Pt: 380 °C (716 °F) |
MDL Number:
MFCD00005005 CAS Number: 89-32-7 EINECS: 201-898-9 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...