Benzyl carbazate ≥98%

Supplier: BeanTown Chemical

Synonyms: Hydrazine, N-CBZ protected, Benzyl hydrazinecarboxylate

145645-100G 145645-25G 145645-5G
BT145645-100GEA 494 USD
BT145645-100G BT145645-25G BT145645-5G
Benzyl carbazate ≥98%
Benzyl carbazate

CAS: 5331-43-1; EC No: 226-230-3; MDL No: MFCD00041890; RTECS: MV1724950 Solid; Molecular Formula: C8H10N2O; MW: 166.18 Melting Point: 65-68°


  • RTECS: MV1724950
  • Solid

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Melting Pt: 65…68 °C
MDL Number: MFCD00041890
CAS Number: 5331-43-1
EINECS: 226-230-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR