Butyrophenone ≥99%

Supplier: BeanTown Chemical

Synonyms: 1-Phenylbutan-1-one, Phenyl propyl ketone, 1-Phenyl-1-butanone

145640-250G 145640-50G
BT145640-250GEA 180 USD
BT145640-250G BT145640-50G
Butyrophenone ≥99%
Butyrophenone

CAS: 495-40-9; EC No: 207-799-7; MDL No: MFCD00009397 Liquid; Molecular Formula: C10H12O; MW: 148.20 Melting Point: 11-13°; Boiling Point: 228-230°; Flash point: 97°C (207°F) Density (g/mL): 1.021; Refractive Index: 1.520


  • Refractive index: 1.520
  • Liquid

Formula: C₆H₅C(O)(CH₂)₂CH₃
MW: 148.2 g/mol
Boiling Pt: 228…230 °C
Melting Pt: 11…13 °C
Density: 1.021 g/mL
Flash Pt: 97 °C (207 °F)
MDL Number: MFCD00009397
CAS Number: 495-40-9
EINECS: 207-799-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74d21a84719c71ec28f51844913f7376027ac053&vid=a8921a2a2e416872169c2c45fe321889cd871e13&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR