Copper(II) pyrophosphate hydrate

Supplier: BeanTown Chemical

Synonyms: Copper(II) diphosphate xhydrate, Cupric pyrophosphate hydrate

145630-1KG 145630-5KG
BT145630-1KGEA 69 USD
BT145630-1KG BT145630-5KG
Copper(II) pyrophosphate hydrate
Copper(II) pyrophosphate hydrate

CAS: 304671-71-4; EC No: 239-250-2; MDL No: MFCD00150225 Powder; Linear Formula: Cu2P2O7·xH2O; MW: 301.02 (anhydrous)


  • Powder

Formula: Cu₂P₂O₇·xH₂O
MW: 301.02 g/mol (anhy)
MDL Number: MFCD00150225
CAS Number: 16570-28-8
EINECS: 239-250-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR