4-Fluoro-3-nitrobenzoic acid ≥98%

Supplier: BeanTown Chemical

145595-100G 145595-25G 145595-5G
BT145595-100GEA 514 USD
BT145595-100G BT145595-25G BT145595-5G
4-Fluoro-3-nitrobenzoic acid ≥98%
4-Fluoro-3-nitrobenzoic acid

CAS: 453-71-4; EC No: 207-221-3; MDL No: MFCD00007058 Solid; Molecular Formula: C7H4FNO4; MW: 185.11 Melting Point: 123-126°


  • Solid

Formula: C₇H₄FNO₄
Melting Pt: 123…126 °C
MDL Number: MFCD00007058
CAS Number: 453-71-4
EINECS: 207-221-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR