Cobalt(II) acetylacetonate

Supplier: BeanTown Chemical

Synonyms: Cobalt(II) bis(acetylacetonate), Cobalt (II) bis(2,4-pentanedionato-O,O'), Cobalt (II) 2,4-pentanedionate, Bis(2,4-pentanedionato-O,O')cobalt (II)

145590-250G 145590-50G
BT145590-250GEA 216.11 USD
BT145590-250G BT145590-50G
Cobalt(II) acetylacetonate
Cobalt(II) acetylacetonate

CAS: 14024-48-7; EC No: 237-855-6; MDL No: MFCD00000014 Crystalline; Molecular Formula: C10H14CoO4 ; MW: 257.14 Melting Point: 165-170° Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: C₁₀H₁₄CoO₄
MW: 257.15 g/mol
Melting Pt: 165…170 °C
MDL Number: MFCD00000014
CAS Number: 14024-48-7
EINECS: 237-855-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR