ε-Hexalactone ≥99%
Supplier: BeanTown Chemical
Synonyms:
ε-Caprolactone, Oxepan-2-one, Hexan-6-olide
145530-100G
145530-2.5KG
145530-500G
BT145530-100GEA
24
USD
BT145530-100G
BT145530-2.5KG
BT145530-500G
ε-Hexalactone ≥99%
ε-Hexalactone
CAS: 502-44-3; EC No: 207-938-1; MDL No: MFCD00003267; RTECS: MO8400000
Liquid; Molecular Formula: C6H10O2; MW: 114.14
Melting Point: -2°; Boiling Point: 235-236°; Flash point: 109°C (228°F)
Density (g/mL): 1.03; Refractive Index: 1.463
- RTECS: MO8400000
- Refractive index: 1.463
- Liquid
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol Boiling Pt: 235…236 °C Melting Pt: –2 °C Density: 1.03 g/mL Flash Pt: 109 °C (228 °F) |
MDL Number:
MFCD00003267 CAS Number: 502-44-3 EINECS: 207-938-1 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...