Bis(2,4-pentanedionato)molybdenum(VI) dioxide ≥99%
Supplier: BeanTown Chemical
Synonyms:
Molybdenum(VI)dioxy Acetylacetonate, Acetylacetone Molybdenum(VI)dioxy Salt, Molybdenum(VI) oxide bis(acetylacetonate), Molybdenum(VI) dioxide bis(acetylacetonate), Molybdenyl acetylacetonate, Bis(acetylacetonato)dioxomolybdenum(VI)
145520-100G
145520-25G
145520-5G
BT145520-100GEA
303
USD
BT145520-100G
BT145520-25G
BT145520-5G
Bis(2,4-pentanedionato)molybdenum(VI) dioxide ≥99%
Bis(2,4-pentanedionato)molybdenum(VI) dioxide
CAS: 17524-05-9; EC No: 241-522-0; MDL No: MFCD00011506
Powder; Linear Formula: Mo(O)2(C5H7O2)2; Molecular Formula: C10H14MoO6; MW: 326.16
Melting Point: 184° (decomposes)
- Powder
Formula:
C₁₀H₁₄MoO₆ MW: 326.18 g/mol Melting Pt: 184 °C (dec.) |
MDL Number:
MFCD00011506 CAS Number: 17524-05-9 EINECS: 241-522-0 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...