Vanillic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Hydroxy-3-methoxybenzoic acid

145505-100G 145505-25G 145505-500G
BT145505-100GEA 115.85 USD
BT145505-100G BT145505-25G BT145505-500G
Vanillic acid ≥98%
Vanillic acid

CAS: 121-34-6; EC No: 204-466-8; MDL No: MFCD00002551; RTECS: YW5300000 Powder; Linear Formula: (HO)C6H3(OCH3)COOH; Molecular Formula: C8H8O4; MW: 168.15 Melting Point: 208-210°


  • RTECS: YW5300000
  • Powder

Formula: C₈H₈O₄
Melting Pt: 208…210 °C
MDL Number: MFCD00002551
CAS Number: 121-34-6
EINECS: 204-466-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR