Rhodium(III) oxide pentahydrate ≥99.99%

Supplier: BeanTown Chemical

145430-1G 145430-250MG 145430-5G
BT145430-1GEA 336 USD
BT145430-1G BT145430-250MG BT145430-5G
Rhodium(III) oxide pentahydrate ≥99.99%
Rhodium (III) oxide pentahydrate

CAS: 39373-27-8; EC No: 234-846-9; MDL No: MFCD00149843 Powder; Linear Formula: Rh2O3·5H2O; MW: 343.88


  • Powder

Formula: O₃Rh₂·5H₂O
MW: 343.88 g/mol (253.81 anhy)
MDL Number: MFCD00149843
CAS Number: 39373-27-8
EINECS: 234-846-9

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR