O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride ≥99%
Supplier: BeanTown Chemical
145415-1G
145415-250MG
145415-5G
BT145415-1GEA
179.17
USD
BT145415-1G
BT145415-250MG
BT145415-5G
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride ≥99%
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride
CAS: 57981-02-9; EC No: 261-057-7; MDL No: MFCD00012953
Solid; Linear Formula: C6F5CH2ONH2·HCl; Molecular Formula: C7H4F5NO·HCl; MW: 249.57
Melting Point: 227° (sublimes)
- Solid
Formula:
C₇H₄F₅NO Melting Pt: 227 °C (subl.) |
MDL Number:
MFCD00012953 CAS Number: 57981-02-9 EINECS: 261-057-7 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...