Tris(dibenzylideneacetone)dipalladium(0) ≥98%
Supplier: BeanTown Chemical
Synonyms:
Pd2(dba)3, Pd2dba3, Tris(dibenzylideneacetonyl)bis-palladium, Bis[tris(dibenzylideneacetone)palladium(0)]
145410-1G
145410-5G
BT145410-1GEA
80.56
USD
BT145410-1G
BT145410-5G
Tris(dibenzylideneacetone)dipalladium(0) ≥98%
Tris(dibenzylideneacetone)dipalladium(0)
CAS: 51364-51-3(52409-22-0); EC No: 479-280-5; MDL No: MFCD00013310
Powder; Linear Formula: (C6H5CH=CHCOCH=CHC6H5)3Pd2; Molecular Formula: C51H42O3Pd2; MW: 915.72
Melting Point: 152-155°
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Powder
Formula:
C₅₁H₄₂O₃Pd₂ MW: 915.72 g/mol Melting Pt: 152…155 °C |
MDL Number:
MFCD00013310 CAS Number: 51364-51-3 EINECS: 479-280-5 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...