Potassium hexacyanoferrate(III) ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide

145375-2KG 145375-500G
BT145375-2KGEA 223 USD
BT145375-2KG BT145375-500G
Potassium hexacyanoferrate(III) ≥99.0% ACS
Potassium hexacyanoferrate(III)

CAS: 13746-66-2; EC No: 237-323-3; MDL No: MFCD00011392; RTECS: LJ8225000 Crystalline; Linear Formula: K3Fe(CN)6; MW: 329.26 Density (g/mL): 1.89 Light Sensitive


  • RTECS: LJ8225000
  • Light Sensitive
  • Crystalline

Formula: K₃Fe(CN)₆
MW: 329.25 g/mol
Density: 1.89 g/mL
MDL Number: MFCD00011392
CAS Number: 13746-66-2
EINECS: 237-323-3

Order Now

Specification Test Results

Assay 99.0% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR