(3-Glycidoxypropyl)trimethoxysilane ≥97%

Supplier: BeanTown Chemical

Synonyms: (3-Glycidyloxypropyl)trimethoxysilane, 3-Glycidoxypropyltrimethoxysilane, (3-(2,3-Epoxypropoxy)propyl)trimethoxysilane

145300-100G 145300-25G 145300-500G
BT145300-100GEA 45.99 USD
BT145300-100G BT145300-25G BT145300-500G
(3-Glycidoxypropyl)trimethoxysilane ≥97%
(3-Glycidoxypropyl)trimethoxysilane

CAS: 2530-83-8; EC No: 219-784-2; MDL No: MFCD00005144; RTECS: VV4025000 Liquid; Molecular Formula: C9H20O5Si; MW: 236.34 Boiling Point: 120°/2 mmHg; Flash point: 122°C (252°F) Density (g/mL): 1.07; Refractive Index: 1.429 Moisture Sensitive


  • RTECS: VV4025000
  • Refractive index: 1.429
  • Moisture Sensitive
  • Liquid

Formula: C₉H₂₀O₅Si
MW: 236.34 g/mol
Boiling Pt: 120 °C/2 mmHg
Density: 1.07 g/mL
Flash Pt: 122 °C (252 °F)
MDL Number: MFCD00005144
CAS Number: 2530-83-8
EINECS: 219-784-2

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR