(S)-(-)-2-Amino-3-methyl-1,1-diphenyl-1-butanol ≥98%

Supplier: BeanTown Chemical

Synonyms: (S)-2-Amino-3-methyl-1,1-diphenylbutan-1-ol

145265-1G 145265-5G
BT145265-1GEA 202 USD
BT145265-1G BT145265-5G
(S)-(-)-2-Amino-3-methyl-1,1-diphenyl-1-butanol ≥98%
(S)-(-)-2-Amino-3-methyl-1,1-diphenyl-1-butanol

CAS: 78603-95-9; MDL No: MFCD01318248 Powder; Linear Formula: (CH3)2CHCH(NH2)C(C6H5)2(OH); Molecular Formula: C17H21NO; MW: 255.35 Melting Point: 95-99° Optical Rotation: [α]20/D -132°, c = 1 in chloroform


  • Optical rotation: [α]20/D -132°, c = 1 in chloroform
  • Powder

Formula: C₁₇H₂₁NO
MW: 255.36 g/mol
Melting Pt: 95…99 °C
MDL Number: MFCD01318248
CAS Number: 78603-95-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR