Sodium hexacyanoferrate(II) decahydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: Sodium ferrocyanide decahydrate

145245-250G 145245-50G
BT145245-250GEA 140 USD
BT145245-250G BT145245-50G
Sodium hexacyanoferrate(II) decahydrate ≥99%
Sodium hexacyanoferrate(II) decahydrate

CAS: 14434-22-1; EC No: 237-081-9; MDL No: MFCD00085360; RTECS: LJ8145000 Powder; Linear Formula: Na4Fe(CN)6·10H2O; Molecular Formula: C6FeN6Na4·10H2O; MW: 484.07 Density (g/mL): 1.458


  • RTECS: LJ8145000
  • Powder

Formula: C₆H₂₀FeN₆Na₄O₁₀
MW: 484.062 g/mol
Density: 1.458 g/mL
MDL Number: MFCD00085360
CAS Number: 14434-22-1
EINECS: 237-081-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR