4-Bromobenzaldehyde ≥98%
Supplier: BeanTown Chemical
145230-100G
145230-25G
145230-500G
BT145230-100GEA
230.02
USD
BT145230-100G
BT145230-25G
BT145230-500G
4-Bromobenzaldehyde ≥98%
4-Bromobenzaldehyde
CAS: 1122-91-4; EC No: 214-365-0; MDL No: MFCD00003377; RTECS: CU4810000
Crystalline; Molecular Formula: C7H5BrO; MW: 185.02
Melting Point: 55-58°; Flash point: 109°C (228°F)
Air Sensitive
- RTECS: CU4810000
- Air Sensitive
- Crystalline
Formula:
C₇H₅BrO Melting Pt: 55…58 °C Flash Pt: 109 °C (228 °F) |
MDL Number:
MFCD00003377 CAS Number: 1122-91-4 EINECS: 214-365-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...