Fumaric acid ≥99%

Supplier: BeanTown Chemical

Synonyms: trans-1,2-Ethylentricarboxylic acid

145210-10KG 145210-2.5KG 145210-500G
BT145210-10KGEA 319 USD
BT145210-10KG BT145210-2.5KG BT145210-500G
Fumaric acid ≥99%
Fumaric acid

CAS: 110-17-8; EC No: 203-743-0; MDL No: MFCD00002700; RTECS: LS9625000 Solid; Linear Formula: HO2CCH=CHCO2H; Molecular Formula: C4H4O4; MW: 116.07 Melting Point: 298-300° (sublimes); Flash point: 230°C (446°F)


  • RTECS: LS9625000
  • Solid

Formula: C₄H₄O₄
MW: 116.07 g/mol
Melting Pt: 298…300 °C (subl.)
Flash Pt: 230 °C (446 °F)
MDL Number: MFCD00002700
CAS Number: 110-17-8
EINECS: 203-743-0

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR