D-(+)-Biotin ≥98%
Supplier: BeanTown Chemical
Synonyms:
Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
145170-10G
145170-1G
145170-5G
BT145170-10GEA
454
USD
BT145170-10G
BT145170-1G
BT145170-5G
D-(+)-Biotin ≥98%
D-(+)-Biotin
CAS: 58-85-5; EC No: 200-399-3; MDL No: MFCD00005541; RTECS: XJ9088200
Powder; Molecular Formula: C10H16N2O3S; MW: 244.32
Melting Point: 229-234°
Optical Rotation: [α]20/D +91±2°, c = 1% in 0.1 M NaOH
Light Sensitive
- RTECS: XJ9088200
- Optical rotation: [α]20/D +91±2°, c = 1% in 0.1 M NaOH
- Light Sensitive
- Powder
Formula:
C₁₀H₁₆N₂O₃S MW: 244.31 g/mol Melting Pt: 229…234 °C |
MDL Number:
MFCD00005541 CAS Number: 58-85-5 EINECS: 200-399-3 Merck Index: 13,01231 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...