3-Amino-1H-1,2,4-triazole-5-thiol ≥98%
Supplier: BeanTown Chemical
Synonyms:
3-Amino-5-mercapto-1,2,4-triazole, 3-Amino-1,2,4-triazole-5-thiol, 3-Amino-4H-1,2,4-triazole-5-thiol, 3-Amino-5-mercapto-1,2,4-triazole 5-Amino-4H-1,2,4-triazole-3-thiol
145165-100G
145165-25G
145165-5G
BT145165-100GEA
366
USD
BT145165-100G
BT145165-25G
BT145165-5G
3-Amino-1H-1,2,4-triazole-5-thiol ≥98%
3-Amino-1H-1,2,4-triazole-5-thiol
CAS: 16691-43-3; EC No: 240-735-6; MDL No: MFCD00005231; RTECS: XZ5357500
Powder; Molecular Formula: C2H4N4; MW: 116.14
Melting Point:
- RTECS: XZ5357500
- Powder
Formula:
C₂H₄N₄S MW: 116.15 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00005231 CAS Number: 16691-43-3 EINECS: 240-735-6 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...