Ethyl acetimidate hydrochloride ≥97%
Supplier: BeanTown Chemical
145140-10G
145140-250G
145140-50G
BT145140-10GEA
21
USD
BT145140-10G
BT145140-250G
BT145140-50G
Ethyl acetimidate hydrochloride ≥97%
Ethyl acetimidate hydrochloride
CAS: 2208-07-3; EC No: 218-631-7; MDL No: MFCD00012572
Powder; Molecular Formula: C4H9NO·HCl; MW: 123.58
Melting Point: 112-114°
Moisture Sensitive
- Moisture Sensitive
- Powder
Formula:
C₄H₁₀ClNO Melting Pt: 112…114 °C |
MDL Number:
MFCD00012572 CAS Number: 2208-07-3 EINECS: 218-631-7 |
Specification Test Results
Assay | 97% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...