N-Methyl-4-nitrophthalimide ≥99%

Supplier: BeanTown Chemical

Synonyms: 2-Methyl-5-nitroisoindoline-1,3-dione

145135-25G 145135-5G
BT145135-25GEA 98 USD
BT145135-25G BT145135-5G
N-Methyl-4-nitrophthalimide ≥99%
N-Methyl-4-nitrophthalimide

CAS: 41663-84-7; EC No: 255-483-2; MDL No: MFCD00454263 ; RTECS: NR3398541 Solid; Molecular Formula: C9H6N2O4; MW: 206.15 Melting Point: 94-98°


  • RTECS: NR3398541
  • Solid

Formula: C₉H₆N₂O₄
MW: 206.16 g/mol
Melting Pt: 94…98 °C
MDL Number: MFCD00454263
CAS Number: 41663-84-7
EINECS: 255-483-2

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR