Cobalt(II) Phthalocyanine
Supplier: BeanTown Chemical
Synonyms:
Cobalt(II) phthalocyanine, Phthalocyanine Cobalt(II), 29H,31H-Phthalocyanine, cobalt complex
145130-1G
145130-5G
BT145130-1GEA
21
USD
BT145130-1G
BT145130-5G
Cobalt(II) Phthalocyanine
Cobalt(II) Phthalocyanine
CAS: 3317-67-7; EC No: 222-012-7; MDL No: MFCD00010718
Powder; Molecular Formula: C32H16CoN8; MW: 571.46
- Powder
Formula:
C₃₂H₁₆CoN₈ MW: 571.47 g/mol |
MDL Number:
MFCD00010718 CAS Number: 3317-67-7 EINECS: 222-012-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...