trans-1-Ethoxy-4-(4-pentylcyclohexyl)benzene ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-(trans-4-Pentylcyclohexyl)-4-ethoxybenzene, 1-Ethoxy-4-(trans-4-amylcyclohexyl)benzene, 1-Ethoxy-4-(trans-4-pentylcyclohexyl)benzene

145125-1G 145125-5G
BT145125-1GEA 103 USD
BT145125-1G BT145125-5G
trans-1-Ethoxy-4-(4-pentylcyclohexyl)benzene ≥97%
trans-1-Ethoxy-4-(4-pentylcyclohexyl)benzene

CAS: 84540-32-9; EC No: 283-125-5; MDL No: MFCD06658196 Solid; Molecular Formula: C19H30O; MW: 274.45 Melting Point: 51-52°


  • Solid

Formula: C₁₉H₃₀O
MW: 274.448 g/mol
Melting Pt: 51…52 °C
MDL Number: MFCD06658196
CAS Number: 84540-32-9
EINECS: 283-125-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR