Potassium hexacyanoferrate(III) ≥98%

Supplier: BeanTown Chemical

Synonyms: Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide

145120-1KG 145120-250G 145120-5KG
BT145120-1KGEA 80.25 USD
BT145120-1KG BT145120-250G BT145120-5KG
Potassium hexacyanoferrate(III) ≥98%
Potassium hexacyanoferrate(III)

CAS: 13746-66-2; EC No: 237-323-3; MDL No: MFCD00011392; RTECS: LJ8225000 Crystalline; Linear Formula: K3Fe(CN)6; MW: 329.26 Density (g/mL): 1.89 Light Sensitive


  • RTECS: LJ8225000
  • Light Sensitive
  • Crystalline

Formula: K₃Fe(CN)₆
MW: 329.25 g/mol
Density: 1.89 g/mL
MDL Number: MFCD00011392
CAS Number: 13746-66-2
EINECS: 237-323-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR