2,6-Dihydroxynaphthalene ≥98%

Supplier: BeanTown Chemical

Synonyms: Naphthalene-2,6-diol

145115-25G 145115-5G
BT145115-25GEA 246 USD
BT145115-25G BT145115-5G
2,6-Dihydroxynaphthalene ≥98%
2,6-Dihydroxynaphthalene

CAS: 581-43-1; EC No: 209-465-6; MDL No: MFCD00004082 Crystalline; Molecular Formula: C10H8O2; MW: 160.17 Melting Point: 223-225°


  • Crystalline

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Melting Pt: 223…225 °C
MDL Number: MFCD00004082
CAS Number: 581-43-1
EINECS: 209-465-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR