2,4,6-Tris(trifluoromethyl)-1,3,5-triazine ≥98%
Supplier: BeanTown Chemical
Synonyms:
2,4,6-Tris(trifluoromethyl)-s-triazine
145095-1G
145095-5G
BT145095-1GEA
35
USD
BT145095-1G
BT145095-5G
2,4,6-Tris(trifluoromethyl)-1,3,5-triazine ≥98%
2,4,6-Tris(trifluoromethyl)-1,3,5-triazine
CAS: 368-66-1; EC No: 206-709-3; MDL No: MFCD00042436; RTECS: XZ2800000
Low Melting Solid; Molecular Formula: C6F9N3 ; MW: 285.07
Boiling Point: 94-99°/748 mmHg
Density (g/mL): 1.596 ; Refractive Index: 1.313
- RTECS: XZ2800000
- Refractive index: 1.313
- Low Melting Solid
Formula:
C₆F₉N₃ Boiling Pt: 94…99 °C/748 mmHg Density: 1.596 g/mL |
MDL Number:
MFCD00042436 CAS Number: 368-66-1 EINECS: 206-709-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...