Lithium acetate dihydrate
Supplier: BeanTown Chemical
Synonyms:
Acetic acid lithium salt
145075-1KG
145075-250G
BT145075-1KGEA
301.65
USD
BT145075-1KG
BT145075-250G
Lithium acetate dihydrate
Lithium acetate dihydrate
CAS: 6108-17-4; EC No: 208-914-3; MDL No: MFCD00066949; RTECS: AI5450000
Crystalline; Linear Formula: LiOOCCH3·2H2O; Molecular Formula: C2H3LiO2·2H2O; MW: 102.01
Melting Point: 53-56°
- RTECS: AI5450000
- Crystalline
Formula:
H₃CCOOLi·2H₂O Melting Pt: 53…56 °C |
MDL Number:
MFCD00066949 CAS Number: 6108-17-4 EINECS: 208-914-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...