Bromophenol blue ACS

Supplier: BeanTown Chemical

Synonyms: Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein

145060-1G 145060-25G 145060-5G
BT145060-1GEA 11.55 USD
BT145060-1G BT145060-25G BT145060-5G
Bromophenol blue ACS
Bromophenol blue

CAS: 115-39-9; EC No: 204-086-2; MDL No: MFCD00005875; RTECS: SJ7453000 Crystalline; Molecular Formula: C19H10Br4O5S; MW: 669.96 Melting Point: 273° (decomposes)


  • RTECS: SJ7453000
  • Crystalline

Formula: C₁₉H₁₀Br₄O₅S
MW: 669.97 g/mol
Melting Pt: 273 °C (dec.)
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR