Europium(III) acetylacetonate hydrate ≥99.9%
Supplier: BeanTown Chemical
145040-1G
145040-5G
BT145040-1GEA
69
USD
BT145040-1G
BT145040-5G
Europium(III) acetylacetonate hydrate ≥99.9%
Europium(III) acetylacetonate hydrate
CAS: 181266-82-0; EC No: 238-185-7; MDL No: MFCD00192170
Powder; Molecular Formula: C15H21EuO6·xH2O ; MW: 449.29 (anhydrous)
Melting Point: 140° (decomposes)
Hygroscopic
- Hygroscopic
- Powder
Formula:
C₁₅H₂₁EuO₆·xH₂O MW: 449.29 g/mol (anhy) Melting Pt: 140 °C (dec.) |
MDL Number:
MFCD00192170 CAS Number: 181266-82-0 EINECS: 238-185-7 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...