Gold(I) sodium thiosulfate hydrate ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

144990-1G 144990-25G 144990-5G
BT144990-1GEA 130 USD
BT144990-1G BT144990-25G BT144990-5G
Gold(I) sodium thiosulfate hydrate ≥99.9% (trace metals basis)
Gold(I) sodium thiosulfate hydrate

CAS: 15283-45-1; EC No: 239-324-4; MDL No: MFCD00046176; RTECS: CK6235000 Powder; Linear Formula: Na3Au(S2O3)2·xH2O; MW: 490.19 (anhydrous)


  • RTECS: CK6235000
  • Powder

Formula: AuH₂Na₃O₆S₄·xH₂O
MDL Number: MFCD00046176
CAS Number: 15283-45-1
EINECS: 239-324-4

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR