(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine ≥99%, ee 98% ee

Supplier: BeanTown Chemical

Synonyms: (R)-1,2,3,4-Tetrahydronaphthalen-1-amine

144975-1G 144975-25G 144975-5G
BT144975-1GEA 37 USD
BT144975-1G BT144975-25G BT144975-5G
(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine ≥99%, ee 98% ee
(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine

CAS: 23357-46-2; MDL No: MFCD00671629 Liquid; Molecular Formula: C10H13N; MW: 147.22 Boiling Point: 118-120°/10 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.002; Refractive Index: 1.565 Air Sensitive


  • Refractive index: 1.565
  • Air Sensitive
  • Liquid

Formula: C₁₀H₁₃N
MW: 147.22 g/mol
Boiling Pt: 118…120 °C/10 mmHg
Density: 1.002 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00671629
CAS Number: 23357-46-2

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR