(R)-(-)-Hydratropic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
(R)-(-)-2-Phenylpropanoic acid, (R)-(-)-2-Phenylpropionic acid, (R)-2-Phenylpropanoic acid
144965-1G
144965-250MG
144965-5G
BT144965-1GEA
176
USD
BT144965-1G
BT144965-250MG
BT144965-5G
(R)-(-)-Hydratropic acid ≥97%
(R)-(-)-Hydratropic acid
CAS: 7782-26-5; MDL No: MFCD00063140
Crystalline; Molecular Formula: C9H10O2 ; MW: 150.17
Melting Point: 29-30°; Boiling Point: 115°/1 mmHg; Flash point: 110°C (230°F)
Density (g/mL): 1.1; Optical Rotation: [α]20/D −72°, c = 1.6 in chloroform
- Optical rotation: [α]20/D −72°, c = 1.6 in chloroform
- Crystalline
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 115 °C/1 mmHg Melting Pt: 29…30 °C Density: 1.1 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00063140 CAS Number: 7782-26-5 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...