3-Amino-9-ethyl carbazole (AEC) ≥95%

Supplier: BeanTown Chemical

Synonyms: 9-Ethylcarbazol-3-ylamine, 9-Ethyl-9H-carbazol-3-amine, AEC, 50X, AEC, 3-Amino-9-ethylcarbazole

144955-100G 144955-25G
BT144955-100GEA 217 USD
BT144955-100G BT144955-25G
3-Amino-9-ethyl carbazole (AEC) ≥95%
3-Amino-9-ethyl carbazole (AEC)

CAS: 132-32-1; EC No: 205-055-7; MDL No: MFCD00004964; RTECS: FE3590000 Powder; Molecular Formula: C14H14N2; MW: 210.27


  • RTECS: FE3590000
  • Powder

Formula: C₁₄H₁₄N₂
MW: 210.28 g/mol
MDL Number: MFCD00004964
CAS Number: 132-32-1
EINECS: 205-055-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR