4-Methoxybenzyl alcohol ≥98%

Supplier: BeanTown Chemical

Synonyms: Anisyl alcohol, p-Anisyl Alcohol, 4-(Hydroxymethyl)anisole, Anise alcohol, (4-Methoxyphenyl)methanol

144945-100G 144945-500G
BT144945-100GEA 51.62 USD
BT144945-100G BT144945-500G
4-Methoxybenzyl alcohol ≥98%
4-Methoxybenzyl alcohol

CAS: 105-13-5; EC No: 203-273-6; MDL No: MFCD00004653; RTECS: DO8925000 Liquid/Low Melting Solid; Molecular Formula: C8H10O2; MW: 138.16 Melting Point: 22-25°; Boiling Point: 259°; Flash point: 110°C (230°F) Density (g/mL): 1.113; Refractive Index: 1.544


  • RTECS: DO8925000
  • Refractive index: 1.544
  • Liquid/Low Melting Solid

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 259 °C
Melting Pt: 22…25 °C
Density: 1.113 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00004653
CAS Number: 105-13-5
EINECS: 203-273-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR